Assay Details
Gene Symbol: lamtor3-like | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142094.1 | NP_001135566.1 | 5 - 6 | 316 | 101 | |
GenBank mRNA | BC168057.1 | - | 5 - 6 | 316 | 101 | |
CU075450.1 | - | 5 - 6 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
