Assay Details
Gene Symbol: shf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142104.1 | NP_001135576.1 | 1 - 2 | 723 | 65 | |
XM_012970180.2 | XP_012825634.1 | 1 - 2 | 1166 | 65 | ||
XM_012970181.2 | XP_012825635.1 | 1 - 2 | 1165 | 65 | ||
XM_012970183.2 | XP_012825637.1 | 1 - 2 | 1165 | 65 | ||
XM_018091966.1 | XP_017947455.1 | 1 - 2 | 1167 | 65 | ||
GenBank mRNA | BC168081.1 | - | 1 - 2 | 723 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
