Assay Details
Gene Symbol: chmp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142136.1 | NP_001135608.1 | 3 - 4 | 207 | 64 | |
GenBank mRNA | BC168427.1 | - | 3 - 4 | 207 | 64 | |
CU075373.1 | - | 3 - 4 | 120 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
