Assay Details
| Gene Symbol: vamp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142150.1 | NP_001135622.1 | 5 - 6 | 473 | 81 | |
| GenBank mRNA | BC168462.1 | - | 5 - 6 | 473 | 81 | |
| BC168463.1 | - | 5 - 6 | 473 | 81 | ||
More Information
Set Membership: |
|

vamp7
Genomic Map