Assay Details
Gene Symbol: ndufaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142201.1 | NP_001135673.1 | 1 - 2 | 141 | 95 | |
GenBank mRNA | BC168591.1 | - | 1 - 2 | 141 | 95 | |
CU024975.1 | - | 1 - 2 | 211 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
