Assay Details
Gene Symbol: ndufaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142201.1 | NP_001135673.1 | 2 - 3 | 243 | 101 | |
GenBank mRNA | BC168591.1 | - | 2 - 3 | 243 | 101 | |
CR942548.2 | - | 2 - 3 | 255 | 101 | ||
CU024975.1 | - | 2 - 3 | 313 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
