Assay Details
Gene Symbol: phr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142249.1 | NP_001135721.1 | 458 | 62 | ||
GenBank mRNA | BC169177.1 | - | 458 | 62 |
Gene Symbol: LOC101733551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094393.1 | XP_017949882.1 | 1 - 2 | 149 | 62 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018094393.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
