Assay Details
Gene Symbol: phr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142249.1 | NP_001135721.1 | 875 | 70 | ||
GenBank mRNA | BC169177.1 | - | 875 | 70 |
Gene Symbol: LOC101733551 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018094393.1 | XP_017949882.1 | 3 - 4 | 487 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
