Assay Details
Gene Symbol: phr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001142249.1 | NP_001135721.1 | 1027 | 89 | ||
GenBank mRNA | BC169177.1 | - | 1027 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
