Assay Details
| Gene Symbol: cd40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145216.1 | NP_001138688.1 | 4 - 5 | 397 | 60 | |
| GenBank mRNA | FJ534591.1 | - | 4 - 5 | 397 | 60 | |
More Information
Set Membership: |
|

cd40
Genomic Map