Assay Details
Gene Symbol: ccnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145376.2 | NP_001138848.1 | 2 - 3 | 231 | 67 | |
GenBank mRNA | BC080491.1 | - | 2 - 3 | 233 | 67 | |
BC158910.1 | - | 2 - 3 | 195 | 67 | ||
CR760967.2 | - | 2 - 3 | 168 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
