Assay Details
Gene Symbol: krt78.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006715.1 | NP_001006716.1 | 4 - 5 | 1077 | 96 | |
GenBank mRNA | BC075443.1 | - | 4 - 5 | 1077 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
