Assay Details
| Gene Symbol: fcn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004935.1 | NP_001004935.1 | 9 | 846 | 107 | |
| XM_012969758.2 | XP_012825212.1 | 8 | 835 | 107 | ||
| GenBank mRNA | BC075410.1 | - | 9 | 846 | 107 | |
| BC091006.1 | - | 7 | 911 | 107 | ||
More Information
Set Membership: |
|

fcn1
Genomic Map