Assay Details
Gene Symbol: b3galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006126.1 | NP_001006126.1 | 2 | 949 | 90 | |
GenBank mRNA | BC075347.1 | - | 2 | 949 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
