Assay Details
| Gene Symbol: sox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007501.1 | NP_001007502.1 | 1 | 856 | 103 | |
| GenBank mRNA | BC079938.1 | - | 1 | 856 | 103 | |
| CT030436.1 | - | 1 | 811 | 103 | ||
More Information
Set Membership: |
|

sox3
Genomic Map