Assay Details
| Gene Symbol: ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015758.1 | NP_001015758.1 | 2 | 351 | 73 | |
| GenBank mRNA | BC089694.1 | - | 2 | 351 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ifitm3| Gene Symbol: ifitm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015758.1 | NP_001015758.1 | 2 | 351 | 73 | |
| GenBank mRNA | BC089694.1 | - | 2 | 351 | 73 | |
Set Membership: |
|