Assay Details
| Gene Symbol: chrac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001015967.2 | NP_001015967.1 | 3 | 552 | 107 | |
| GenBank mRNA | BC159088.1 | - | 3 | 527 | 107 | |
| CR848420.2 | - | 3 | 552 | 107 | ||
| CR926252.1 | - | 3 | 532 | 107 | ||
More Information
Set Membership: |
|

chrac1
Genomic Map