Assay Details
Gene Symbol: tob2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016213.2 | NP_001016213.1 | 2 | 953 | 76 | |
GenBank mRNA | BC136152.1 | - | 2 | 956 | 76 | |
CR926324.2 | - | 2 | 953 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
