Assay Details
| Gene Symbol: vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016285.3 | NP_001016285.2 | 3 | 657 | 107 | |
| GenBank mRNA | CR761599.2 | - | 3 | 657 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

vamp8| Gene Symbol: vamp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016285.3 | NP_001016285.2 | 3 | 657 | 107 | |
| GenBank mRNA | CR761599.2 | - | 3 | 657 | 107 | |
Set Membership: |
|