Assay Details
Gene Symbol: bud31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016359.3 | NP_001016359.2 | 3 | 184 | 74 | |
XM_012970415.2 | XP_012825869.1 | 3 | 462 | 74 | ||
XM_018097105.1 | XP_017952594.1 | 3 | 161 | 74 | ||
GenBank mRNA | BC135529.1 | - | 3 | 184 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
