Assay Details
Gene Symbol: mphosph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016703.2 | NP_001016703.1 | 5 | 454 | 101 | |
GenBank mRNA | BC159060.1 | - | 5 | 461 | 101 | |
BC170690.1 | - | 5 | 451 | 101 | ||
BC170692.1 | - | 5 | 451 | 101 | ||
CR760141.2 | - | 5 | 454 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
