Assay Details
Gene Symbol: fzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016741.2 | NP_001016741.1 | 1 | 956 | 89 | |
GenBank mRNA | BC090579.1 | - | 1 | 913 | 89 | |
CR759995.2 | - | 1 | 956 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
