Assay Details
| Gene Symbol: sox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016853.2 | NP_001016853.1 | 3 | 1555 | 95 | |
| GenBank mRNA | CR855424.2 | - | 3 | 1555 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

sox9| Gene Symbol: sox9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001016853.2 | NP_001016853.1 | 3 | 1555 | 95 | |
| GenBank mRNA | CR855424.2 | - | 3 | 1555 | 95 | |
Set Membership: |
|