Assay Details
Gene Symbol: cda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017217.2 | NP_001017217.1 | 254 | 86 | ||
GenBank mRNA | BC118900.1 | - | 215 | 86 | ||
CR760377.2 | - | 254 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
