Assay Details
| Gene Symbol: nat14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045600.1 | NP_001039065.1 | 3 | 851 | 68 | |
| XM_012966692.2 | XP_012822146.1 | 4 | 809 | 68 | ||
| XM_012966693.2 | XP_012822147.1 | 4 | 902 | 68 | ||
| XM_012966694.2 | XP_012822148.1 | 3 | 740 | 68 | ||
| XM_012966695.2 | XP_012822149.1 | 3 | 658 | 68 | ||
| XM_012966696.2 | XP_012822150.1 | 3 | 655 | 68 | ||
| XM_018095419.1 | XP_017950908.1 | 4 | 817 | 68 | ||
| GenBank mRNA | CR855496.2 | - | 3 | 851 | 68 | |
More Information
Set Membership: |
|

nat14
Genomic Map