Assay Details
Gene Symbol: rbm8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045682.1 | NP_001039147.1 | 750 | 78 | ||
GenBank mRNA | BC157757.1 | - | 821 | 78 | ||
CR942553.2 | - | 750 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
