Assay Details
| Gene Symbol: akr1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078713.1 | NP_001072181.1 | 1155 | 113 | ||
| GenBank mRNA | BC080355.1 | - | 1155 | 113 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

akr1c3| Gene Symbol: akr1c3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078713.1 | NP_001072181.1 | 1155 | 113 | ||
| GenBank mRNA | BC080355.1 | - | 1155 | 113 | ||
Set Membership: |
|