Assay Details
| Gene Symbol: dusp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078725.1 | NP_001072193.1 | 2 | 955 | 96 | |
| XM_012954136.2 | XP_012809590.1 | 3 | 1097 | 96 | ||
| XM_012954138.2 | XP_012809592.1 | 2 | 1010 | 96 | ||
| XM_012954139.2 | XP_012809593.1 | 3 | 1056 | 96 | ||
| XM_018089971.1 | XP_017945460.1 | 3 | 816 | 96 | ||
| GenBank mRNA | BC118734.1 | - | 2 | 955 | 96 | |
More Information
Set Membership: |
|

dusp14
Genomic Map