Assay Details
| Gene Symbol: tubb4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078982.2 | NP_001072450.1 | 3 | 253 | 99 | |
| XM_012969765.2 | XP_012825219.1 | 2 | 374 | 99 | ||
| GenBank mRNA | BC121707.1 | - | 3 | 239 | 99 | |
| BC135706.1 | - | 3 | 253 | 99 | ||
More Information
Set Membership: |
|

tubb4b
Genomic Map