Assay Details
| Gene Symbol: gja4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079436.1 | NP_001072904.1 | 2 | 1052 | 111 | |
| GenBank mRNA | BC121698.1 | - | 2 | 1052 | 111 | |
More Information
Set Membership: |
|

gja4
Genomic Map