Assay Details
Gene Symbol: bcat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102757.1 | NP_001096227.1 | 755 | 90 | ||
GenBank mRNA | BC121908.1 | - | 755 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
