Assay Details
| Gene Symbol: rpp25l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113880.1 | NP_001107352.1 | 554 | 110 | ||
| GenBank mRNA | BC157189.1 | - | 554 | 110 | ||
| BC171238.1 | - | 517 | 110 | |||
| BC171244.1 | - | 517 | 110 | |||
More Information
Set Membership: |
|

rpp25l
Genomic Map