Assay Details
| Gene Symbol: LOC100135213 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113914.1 | NP_001107386.1 | 2 | 1346 | 180 | |
| Gene Symbol: znf33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC157283.1 | - | 2 | 1346 | 180 | |
More Information
Set Membership: |
|

LOC100135213
Genomic Map