Assay Details
| Gene Symbol: hist1h2ah | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001126645.1 | NP_001120117.1 | 1 | 1054 | 84 | |
| GenBank mRNA | BC158988.1 | - | 1 | 1054 | 84 | |
More Information
Set Membership: |
|

hist1h2ah
Genomic Map