Assay Details
| Gene Symbol: nanos1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203526.1 | NP_988857.1 | 1 | 143 | 91 | |
| GenBank mRNA | AF256086.1 | - | 1 | 143 | 91 | |
| BC160384.1 | - | 1 | 141 | 91 | ||
More Information
Set Membership: |
|

nanos1
Genomic Map