Assay Details
| Gene Symbol: kcnrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203532.1 | NP_988863.1 | 2 | 755 | 78 | |
| GenBank mRNA | AY190924.1 | - | 2 | 755 | 78 | |
| BC074581.1 | - | 2 | 751 | 78 | ||
More Information
Set Membership: |
|

kcnrg
Genomic Map