Assay Details
Gene Symbol: scfd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203735.1 | NP_989066.1 | 1 | 352 | 103 | |
GenBank mRNA | BC061300.1 | - | 1 | 352 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
