Assay Details
| Gene Symbol: drap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203941.1 | NP_989272.1 | 3 | 346 | 105 | |
| GenBank mRNA | BC064181.1 | - | 3 | 346 | 105 | |
| CR761421.2 | - | 3 | 302 | 105 | ||
More Information
Set Membership: |
|

drap1
Genomic Map