Assay Details
| Gene Symbol: arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_204081.1 | NP_989412.1 | 1 | 350 | 92 | |
| XM_012968173.2 | XP_012823627.1 | 2 | 576 | 92 | ||
| GenBank mRNA | BC064861.1 | - | 1 | 350 | 92 | |
| CR942350.2 | - | 2 | 536 | 92 | ||
More Information
Set Membership: |
|

arf6
Genomic Map