Assay Details
Gene Symbol: LOC100274124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001148502.1 | NP_001141974.1 | 1065 | 83 | ||
GenBank mRNA | BT042400.1 | - | 1065 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
