Assay Details
| Gene Symbol: LOC100281116 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154035.1 | NP_001147507.1 | 1657 | 90 | ||
| GenBank mRNA | EU955640.1 | - | 1657 | 90 | ||
More Information
Set Membership: |
|

LOC100281116
Genomic Map