Assay Details
Gene Symbol: LOC100281116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001154035.1 | NP_001147507.1 | 1041 | 97 | ||
GenBank mRNA | EU955640.1 | - | 1041 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
