Assay Details
Gene Symbol: LOC100282249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155161.1 | NP_001148633.1 | 444 | 77 | ||
GenBank mRNA | EU960198.1 | - | 444 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
