Assay Details
| Gene Symbol: LOC100283073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155975.1 | NP_001149447.1 | 915 | 108 | ||
| GenBank mRNA | EU963361.1 | - | 915 | 108 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283073| Gene Symbol: LOC100283073 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155975.1 | NP_001149447.1 | 915 | 108 | ||
| GenBank mRNA | EU963361.1 | - | 915 | 108 | ||