Assay Details
Gene Symbol: LOC100283174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156076.1 | NP_001149548.1 | 999 | 97 | ||
GenBank mRNA | EU963677.1 | - | 999 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
