Assay Details
Gene Symbol: LOC100283932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156830.1 | NP_001150302.1 | 1100 | 93 | ||
GenBank mRNA | BT041229.1 | - | 990 | 93 | ||
EU966462.1 | - | 1100 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
