Assay Details
Gene Symbol: LOC100284061 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001156959.1 | NP_001150431.1 | 1152 | 93 | ||
GenBank mRNA | EU966955.1 | - | 1152 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
