Assay Details
| Gene Symbol: LOC100284118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157015.1 | NP_001150487.1 | 217 | 100 | ||
| GenBank mRNA | BT038766.1 | - | 217 | 100 | ||
| BT054479.1 | - | 126 | 100 | |||
| BT060497.1 | - | 180 | 100 | |||
| EU967137.1 | - | 326 | 100 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT054479.1, EU967137.1 transcripts.
Set Membership: |
|

LOC100284118
Genomic Map