Assay Details
Gene Symbol: LOC100284118 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001157015.1 | NP_001150487.1 | 267 | 94 | ||
GenBank mRNA | BT038766.1 | - | 267 | 94 | ||
BT054479.1 | - | 176 | 94 | |||
BT060497.1 | - | 230 | 94 | |||
EU967137.1 | - | 376 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BT054479.1, EU967137.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
