Assay Details
| Gene Symbol: LOC100284118 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157015.1 | NP_001150487.1 | 304 | 101 | ||
| GenBank mRNA | BT038766.1 | - | 304 | 101 | ||
| BT054479.1 | - | 213 | 101 | |||
| BT060497.1 | - | 267 | 101 | |||
| EU967137.1 | - | 413 | 101 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT054479.1, EU967137.1 transcripts.
Set Membership: |
|

LOC100284118
Genomic Map